PLoS ONE
Investigation of Yersinia pestis Laboratory Adaptation through a Combined Genomics and Proteomics Approach.
PMID 26599979
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6250
2-Mercaptoethanol, ≥99.0%
C2H6OS
M3148
2-Mercaptoethanol, for molecular biology, for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
C2H6OS
M7154
2-Mercaptoethanol, for electrophoresis
C2H6OS
63689
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
C2H6OS
271004
Acetonitrile, anhydrous, 99.8%
C2H3N
675415
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
C2H3N
437557
Acetonitrile, ≥99.5%, ACS reagent
C2H3N
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
639133
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
D8284
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
C20H10N2Na2O4 · xH2O
27002
Formic acid, purum, ≥85%
CH2O2
W248703
Formic acid, ≥95%, FCC, FG
CH2O2
322415
Methanol, anhydrous, 99.8%
CH4O
M1770
Methanol, BioReagent, suitable for protein sequencing
CH4O
650609
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
CH4O
270474
Methanol, for HPLC, gradient grade, 99.93%
CH4O
65548
Methanol, for HPLC, gradient grade, ≥99.8% (GC)
CH4O