EMAIL THIS PAGE TO A FRIEND
Tuberculosis (Edinburgh, Scotland)
The implication of pro-inflammatory cytokines in the impaired production of gonadal androgens by patients with pulmonary tuberculosis.
PMID 26602224