EMAIL THIS PAGE TO A FRIEND
Scientific Reports
An unexpected role for mixotrophs in the response of peatland carbon cycling to climate warming.
PMID 26603894
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5011
5′-Deoxy-5′-(methylthio)adenosine
C11H15N5O3S