EMAIL THIS PAGE TO A FRIEND
Scientific reports
Three-dimensional direct cell patterning in collagen hydrogels with near-infrared femtosecond laser.
PMID 26603915
Abstract
We report a methodology for three-dimensional (3D) cell patterning in a hydrogel in situ. Gold nanorods within a cell-encapsulating collagen hydrogel absorb a focused near-infrared femtosecond laser beam, locally denaturing the collagen and forming channels, into which cells migrate, proliferate, and align in 3D. Importantly, pattern resolution is tunable based on writing speed and laser power, and high cell viability (>90%) is achieved using higher writing speeds and lower laser intensities. Overall, this patterning technique presents a flexible direct-write method that is applicable in tissue engineering systems where 3D alignment is critical (such as vascular, neural, cardiac, and muscle tissue).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ARK2188
Hexadecyltrimethylammonium bromide, PharmaGrade, USP/NF, Manufactured under appropriate controls for pharma or biopharmaceutical production.
C19H42BrN
H9151
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
C19H42BrN
52365
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
C19H42BrN
H6269
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
C19H42BrN
H5882
Hexadecyltrimethylammonium bromide, ≥98%
C19H42BrN
A5960
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
C6H8O6
A7506
L-Ascorbic acid, reagent grade, crystalline
C6H8O6
A0278
L-Ascorbic acid, reagent grade
C6H8O6
A4403
L-Ascorbic acid, powder, cell culture tested, γ-irradiated
C6H8O6
A4544
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
C6H8O6
A2218
L-Ascorbic acid, meets USP testing specifications
C6H8O6
A2174
L-Ascorbic acid, plant cell culture tested
C6H8O6
95209
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
C6H8O6
33034
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.7-100.5% (oxidimetric)
C6H8O6
95210
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
C6H8O6
47863
L-Ascorbic acid, analytical standard
C6H8O6
255564
L-Ascorbic acid, ACS reagent, ≥99%
C6H8O6
A92902
L-Ascorbic acid, 99%
C6H8O6
W210901
L-Ascorbic acid, FCC, FG
C6H8O6
71320
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
H4BNa
452890
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
H4BNa
480886
Sodium borohydride, granular, 99.99% trace metals basis
H4BNa
452874
Sodium borohydride, granular, 10-40 mesh, 98%
H4BNa
452882
Sodium borohydride, powder, ≥98.0%
H4BNa
247677
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
H4BNa
71318
Sodium borohydride, purum, ≥97.0% (gas-volumetric), tablet
H4BNa
213462
Sodium borohydride, ReagentPlus®, 99%
H4BNa
686018
Sodium borohydride, hydrogen-storage grade, 98%
H4BNa
200972
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
H4BNa
452904
Sodium borohydride solution, SWS, ~12 wt. % in 14 M NaOH
H4BNa