EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of nitroalkanes in mainstream cigarette smoke by heart-cutting multidimensional gas chromatography system coupled with mass spectrometry detection.
PMID 26603996
Abstract
In this paper, heart-cutting two-dimensional GC/MS (GC-GC/MS) method in combination with a simple sample collection procedure was developed for the determination of 6 nitroalkanes in mainstream cigarette smoke. The method could remove large amounts of impurities on-line in the first polar column by heart-cuts and separate from the left interferences in a second mid-polar column. And the target compounds could be focused at the inlet of the second column by cryo-concentration. Compared to conventional GC/MS, GC-GC/MS achieved a lower noise level and sensitivity at least an order of magnitude higher. Furthermore, the GC-GC/MS method could avoid the false negative and false positive results that appeared in the compared conventional GC/MS analysis. By trapping the vapor phase of 20 cigarettes smoke, the LODs and LOQs of the nitroalkanes were 1.3 to 9.8 and 4.3 to 32.6ng/cigarette, respectively, and all linear correlation efficiencies were larger than 0.999. The validation results also indicate that the method has high accuracy (spiked recoveries between 84% and 102%) and good repeatability (RSD between 7.2% and 9.4%). The developed method was applied to analyze 1 Kentucky reference cigarette (3R4F) and 10 Chinese commercial brands of cigarettes. The research results indicated that nitromethane, nitroethane, 2-nitropropane and 1-nitro-n-pentane were detected in mainstream cigarette smoke, but 1-nitro-n-butane and 2-nitropropane, which were reported by one previous study, were not detected in all cigarette samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130265
2-Nitropropane, ≥96%
C3H7NO2