EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Synthesis of Plasmonic Cu2-x Se@ZnS Core@Shell Nanoparticles.
PMID 26604183
Abstract
We report the synthesis of plasmonic Cu2-x Se@ZnS core@shell nanoparticles (NPs). We used a shell growth approach, starting from Cu2-x Se NPs that have been shown before to exhibit a localized surface plasmon resonance (LSPR). By careful synthesis planning we avoided cation exchange reactions and received core@shell nanoparticles that, after oxidation under air, exhibit a strong LSPR in the NIR. Interestingly, the crystalline, closed ZnS shell that we grew with variable thickness still allowed a slow oxidation of the core under ambient conditions, while the core was effectively protected from reduction, even in the presence of reducing agents such as borane tert-butyamine complex and diisobutylaluminum hydride, giving rise to a stable particle LSPR, also under strongly reducing conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O806
1-Octadecene, technical grade, 90%
C18H36
256811
Diisobutylaluminum hydride, reagent grade
C8H19Al