PloS one
Long-Acting Beta Agonists Enhance Allergic Airway Disease.
PMID 26605551
Abstract
Asthma is one of the most common of medical illnesses and is treated in part by drugs that activate the beta-2-adrenoceptor (β2-AR) to dilate obstructed airways. Such drugs include long acting beta agonists (LABAs) that are paradoxically linked to excess asthma-related mortality. Here we show that LABAs such as salmeterol and structurally related β2-AR drugs such as formoterol and carvedilol, but not short-acting agonists (SABAs) such as albuterol, promote exaggerated asthma-like allergic airway disease and enhanced airway constriction in mice. We demonstrate that salmeterol aberrantly promotes activation of the allergic disease-related transcription factor signal transducer and activator of transcription 6 (STAT6) in multiple mouse and human cells. A novel inhibitor of STAT6, PM-242H, inhibited initiation of allergic disease induced by airway fungal challenge, reversed established allergic airway disease in mice, and blocked salmeterol-dependent enhanced allergic airway disease. Thus, structurally related β2-AR ligands aberrantly activate STAT6 and promote allergic airway disease. This untoward pharmacological property likely explains adverse outcomes observed with LABAs, which may be overcome by agents that antagonize STAT6.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1263
1,2-Didodecanoyl-sn-glycero-3-phosphocholine, ≥99%, synthetic
C32H64NO8P
D8284
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
C20H10N2Na2O4 · xH2O
C3993
Carvedilol, ≥98% (HPLC), solid
C24H26N2O4
C0378
Chloramphenicol, ≥98% (HPLC)
C11H12Cl2N2O5
C7795
Chloramphenicol, γ-irradiated
C11H12Cl2N2O5
C1919
Chloramphenicol, BioReagent, suitable for plant cell culture
C11H12Cl2N2O5
C0857
Chloramphenicol, meets USP testing specifications
C11H12Cl2N2O5
C1863
Chloramphenicol, Biotechnology Performance Certified, suitable for plant cell culture
C11H12Cl2N2O5
SRP3330 IL-6 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
SRP4145 IL-6 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC)
SRP3096 IL-6 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I9646
Interleukin-6 from mouse, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, carrier free
I0406 Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, cell culture tested
I2786
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, Animal Component Free
I1395
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
H7416 Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
395B
Periodic Acid-Schiff (PAS) Kit
S8260
Salbutamol
C13H21NO3