PLoS ONE
The Nature of Iron Deposits Differs between Symptomatic and Asymptomatic Carotid Atherosclerotic Plaques.
PMID 26606178
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8775
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
CH2O
F8923
Formaldehyde solution, meets USP testing specifications, contains 9.0-15% methanol as stabilizer
CH2O
252549
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
CH2O
F15587
Formaldehyde solution, 37 wt. % in H2O
CH2O
47608
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
CH2O
33220
Formaldehyde solution, puriss. p.a., ACS reagent, ≥36.5%
CH2O
15512
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
CH2O
47083-U
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
CH2O
606758
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
12CH2O
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
435570
Hydrochloric acid, reagent grade, 37%
HCl
40233
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17863), ≥32%
HCl
40253
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
84434
Hydrochloric acid solution, BioXtra, ~0.1 M in H2O
HCl
H0636
Hydrochloric acid solution, protein sequencing grade, liquid
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl