EMAIL THIS PAGE TO A FRIEND
Scientific reports
A chimeolysin with extended-spectrum streptococcal host range found by an induced lysis-based rapid screening method.
PMID 26607832
Abstract
The increasing emergence of multi-drug resistant streptococci poses a serious threat to public health worldwide. Bacteriophage lysins are promising alternatives to antibiotics; however, their narrow lytic spectrum restricted to closely related species is a central shortcoming to their translational development. Here, we describe an efficient method for rapid screening of engineered chimeric lysins and report a unique "chimeolysin", ClyR, with robust activity and an extended-spectrum streptococcal host range against most streptococcal species, including S. pyogenes, S. agalactiae, S. dysgalactiae, S. equi, S. mutans, S. pneumoniae, S. suis and S. uberis, as well as representative enterococcal and staphylococcal species (including MRSA and VISA). ClyR is the first lysin that demonstrates activity against the dominant dental caries-causing pathogen as well as the first lysin that kills all four of the bovine mastitis-causing pathogens. This study demonstrates the success of the screening method resulting in a powerful lysin with potential for treating most streptococcal associated infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5670
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
CaCl2
429759
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.99% trace metals basis
CaCl2
499609
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
CaCl2
C8106
Calcium chloride, meets USP testing specifications
CaCl2 · 2H2O
449709
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
CaCl2
21115
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
CaCl2
C-045
Cannabidiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C21H30O2
2107
E-Toxate Water, endotoxin, free
H2O
I6758
IPTG, ≥99% (TLC), ≤0.1% Dioxane
C9H18O5S
PHG0010
Isopropyl β-D-1-thiogalactopyranoside, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C9H18O5S
I5502
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
C9H18O5S
I1284
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
C9H18O5S
A3256
L-(+)-Arabinose, ≥99%
C5H10O5
A91906
L-(+)-Arabinose, 98%
C5H10O5
10839
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
C5H10O5
W325501
L-(+)-Arabinose, 99%
C5H10O5
S6546
Sodium chloride solution, 5 M
ClNa
16818
Sodium chloride solution, 2 M
ClNa
S8776
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
ClNa
451940
Sodium chloride-35Cl, 99 atom % 35Cl
35ClNa
W4502
Water, Molecular Biology Reagent
H2O
W3500
Water, sterile-filtered, BioReagent, suitable for cell culture
H2O
W1503
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo
H2O
W1754
Water, PCR Reagent
H2O
W3513
Water, BioPerformance Certified
H2O
95289
Water, for cell biology, free of endotoxins, ultrafiltered and autoclaved
H2O
95284
Water, for molecular biology, DEPC-treated and sterile filtered
H2O
697125
Water, packaged for use in deposition systems
H2O
195294
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
H2O
329886
Water-16O, 99.99 atom % 16O
H216O