EMAIL THIS PAGE TO A FRIEND
American Journal of Cancer Research
Up regulation of the long non-coding RNA NEAT1 promotes esophageal squamous cell carcinoma cell progression and correlates with poor prognosis.
PMID 26609486