EMAIL THIS PAGE TO A FRIEND
BioMed Research International
Intake of Meals Containing High Levels of Carbohydrates or High Levels of Unsaturated Fatty Acids Induces Postprandial Dysmetabolism in Young Overweight/Obese Men.
PMID 26609520