EMAIL THIS PAGE TO A FRIEND
BioMed research international
Synergistic Effects of BMP9 and miR-548d-5p on Promoting Osteogenic Differentiation of Mesenchymal Stem Cells.
PMID 26609524
Abstract
Various stimulators have been reported to promote MSC osteogenic differentiation via different pathways such as bone morphogenetic protein 9 (BMP9) through influencing COX-2 and miR-548d-5p through targeting peroxisome proliferator-activated receptor-γ (PPARγ). Whether synergistic effects between BMP9 and miR-548d-5p existed in promoting osteogenesis from MSCs was unclear. In the study, the potential synergistic effects of BMP9 and miR-548d-5p on human MSC differentiation were investigated. Osteogenic differentiation of MSCs treated with BMP9 or miR-548d-5p was detected with multimodality of methods. The results demonstrated that BMP9 and miR-548d-5p significantly influenced COX-2 and PPARγ, respectively. BMP9 also influenced the expression of PPARγ, but no significant effect of miR-548d-5p on COX-2 was observed. When BMP9 and miR-548d-5p were combined, more potent effects on both COX-2 and PPARγ were observed than BMP9 or miR-548d-5p alone. Consistently, osteogenic analysis at different timepoints demonstrated that osteogenic genes, ALP activity, calcium deposition, OPN protein, and matrix mineralization were remarkably upregulated by BMP9/miR-548d-5p compared with BMP9 or miR-548d-5p alone, indicating the synergetic effects of BMP9 and miR-548d-5p on osteogenic differentiation of MSCs. Our study demonstrated that regulating different osteogenic regulators may be an effective strategy to promote bone tissue regeneration for bone defects.