EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Fast pyrolysis char - Assessment of alternative uses within the bioliq® concept.
PMID 26609947
Abstract
Experiments with a process development unit for fast pyrolysis of biomass residues of 10kgh(-1) have been performed to quantify the impact of two different product recovery options. Wheat straw, miscanthus and scrap wood have been used as feedstock. A separate recovery of char increases the organic oil yield as compared to a combined recovery of char and organic condensate (OC). Furthermore, it allows for an alternative use of the byproduct char which represents an important product fraction for the high ash biomass residues under consideration. The char produced shows little advantage over its biomass precursor when considered as energy carrier due to its high ash content. Significant value can be added by demineralizing and activating the char. The potential to increase the economic feasibility of fast pyrolysis is shown by an assessment of the bioliq® process chain.