EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Optical and Electrical Properties of TTF-MPcs (M = Cu, Zn) Interfaces for Optoelectronic Applications.
PMID 26610466
Abstract
Sandwich structures were fabricated by a vacuum deposition method using MPc (M = Cu, Zn), with a Tetrathiafulvalene (TTF) derivative, and Indium Tin Oxide (ITO) and aluminum electrodes. The structure and morphology of the deposited films were studied by IR spectroscopy, scanning electron microscopy (SEM) and energy dispersive spectroscopy (EDS). The absorption spectra of TTF derivative-MPc (M = Cu, Zn) thin films deposited at room temperature were recorded in the spectral range 200-1000 nm. The optical band gap of the thin films was determined from the (αhν)(1/2) vs. hν plot. The direct-current (DC) electrical properties of the glass/ITO/TTFderiv-MPc (M = Cu, Zn)/Al structures were also investigated. Changes in conductivity of the derivative-TTF-enriched Pc compounds suggest the formation of alternative paths for carrier conduction. At low voltages, forward current density obeys an ohmic I-V relationship; at higher voltages, conduction is mostly due to a space-charge-limited conduction (SCLC) mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
695637
Dibenzotetrathiafulvalene, 97%
C14H8S4
183180
Tetrathiafulvalene, 97%
C6H4S4