EMAIL THIS PAGE TO A FRIEND
Scientific reports
Formaldehyde scavengers function as novel antigen retrieval agents.
PMID 26612041
Abstract
Antigen retrieval agents improve the detection of formaldehyde-fixed proteins, but how they work is not well understood. We demonstrate that formaldehyde scavenging represents a key characteristic associated with effective antigen retrieval; under controlled temperature and pH conditions, scavenging improves the typical antigen retrieval process through reversal of formaldehyde-protein adduct formation. This approach provides a rational framework for the identification and development of more effective antigen retrieval agents.