EMAIL THIS PAGE TO A FRIEND
Asian Pacific Journal of Tropical Medicine
Roots extracts of Adenophora triphylla var. japonica improve obesity in 3T3-L1 adipocytes and high-fat diet-induced obese mice.
PMID 26614988