EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Targeting HIF-1α is a prerequisite for cell sensitivity to dichloroacetate (DCA) and metformin.
PMID 26616058
Abstract
Recently, targeting deregulated energy metabolism is an emerging strategy for cancer therapy. In the present study, combination of DCA and metformin markedly induced cell death, compared with each drug alone. Furthermore, the expression levels of glycolytic enzymes including HK2, LDHA and ENO1 were downregulated by two drugs. Interestingly, HIF-1α activation markedly suppressed DCA/metformin-induced cell death and recovered the expressions of glycolytic enzymes that were decreased by two drugs. Based on these findings, we propose that targeting HIF-1α is necessary for cancer metabolism targeted therapy.