EMAIL THIS PAGE TO A FRIEND
Scientific Reports
QUANTITY: An Isobaric Tag for Quantitative Glycomics.
PMID 26616285
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284505
1,2-Dichloroethane, anhydrous, 99.8%
C2H4Cl2
319929
1,2-Dichloroethane, ACS reagent, ≥99.0%
C2H4Cl2
07-2101
1,2-Dichloroethane, for HPLC
C2H4Cl2
85707
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
C7H6O4
149357
2,5-Dihydroxybenzoic acid, 98%
C7H6O4
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
435570
Hydrochloric acid, reagent grade, 37%
HCl
40233
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17863), ≥32%
HCl
40253
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
84434
Hydrochloric acid solution, BioXtra, ~0.1 M in H2O
HCl
H0636
Hydrochloric acid solution, protein sequencing grade, liquid
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
G3126
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
C5H10N2O3
G5792
L-Glutamine, PharmaGrade, Ajinomoto, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H10N2O3
G8540
L-Glutamine, meets USP testing specifications, cell culture tested, 99.0-101.0%, from non-animal source
C5H10N2O3
G6392
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
C5H10N2O3
49419
L-Glutamine, BioUltra, ≥99.5% (NT)
C5H10N2O3
W368401
L-Glutamine
C5H10N2O3
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
D145750
N,N-Dimethylaniline, 99%
C8H11N
407275
N,N-Dimethylaniline, purified by redistillation, ≥99.5%
C8H11N
515124
N,N-Dimethylaniline, ReagentPlus®, 99%
C8H11N
236314
p-Toluidine, 99.6%
C7H9N
461121
p-Toluidine, 99%
C7H9N