EMAIL THIS PAGE TO A FRIEND
Food chemistry
Controlling the taste receptor accessible structure of rebaudioside A via binding to bovine serum albumin.
PMID 26616927
Abstract
We illustrate a method that uses bovine serum albumin (BSA) to control the receptor-accessible part of rebaudioside A (Reb A). The critical micelle concentration (CMC) of Reb A was found to be 4.5 mM and 5 mM at pH 3 and 6.7 respectively. NMR studies show that below its CMC, Reb A binds weakly to BSA to generate a Reb A-protein complex ("RPC"), which is only modestly stable under varying conditions of pH (3.0-6.7) and temperature (4-40°C) with its binding affinities determined to be in the range of 5-280 mM. Furthermore, saturation transfer difference (STD) NMR experiments confirm that the RPC has fast exchange of the bitterness-instigating diterpene of Reb A into the binding sites of BSA. Our method can be used to alter the strength of Reb A-receptor interaction, as a result of binding of Reb A to BSA, which may ultimately lead to moderation of its taste.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0202
DPH, ≥98% (HPLC)
C18H13FN4O2
329916
Potassium dideuterium phosphate, 98 atom % D
D2KO4P