EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Long noncoding RNA AK056155 involved in the development of Loeys-Dietz syndrome through AKT/PI3K signaling pathway.
PMID 26617788
Abstract
Loeys-Dietz syndrome (LDS) is an autosomal dominant genetic connective tissue disorder, and most of LDS patients will develop into aortic aneurysm. Unfortunately, there is no known cure, and a high risk of death from aortic aneurysm rupture. However the detailed mechanism is still unknown. In order to explore the mechanism, we firstly used bioinformatics to predict, and then verified with biology methods. Firstly, we found that LncRNA AK056155 was differentially expressed in peripheral blood circulating endothelial cells between normal patients and LDS patients by bioinformatics. Then we further verified that AK056155 was also overexpressed in aortic aneurysm patients by RT-PCR. Moreover, we demonstrated that the expression of AK056155 can be enhanced by TGF-β1 in a concentration or time depended manner in HUVECs by RT-PCR. Furthermore, the expression of AK056155 was reduced with treatment of PI3K inhibitor (LY294002) or AKT inhibitor (GDC-0068) in combination with TGF-β1. These results indicate that AK056155 involved in the development of Loeys-Dietz syndrome through AKT/PI3K signaling pathway, it may provide a promising target gene to prevent LDS develop in to aortic aneurysm.