EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Acetylation of glucokinase regulatory protein decreases glucose metabolism by suppressing glucokinase activity.
PMID 26620281
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8375
2-Deoxy-D-glucose, ≥98% (GC), crystalline
C6H12O5
D6134
2-Deoxy-D-glucose, ≥99% (GC), crystalline
C6H12O5
D3179
2-Deoxy-D-glucose, BioXtra, ≥98%
C6H12O5
271004
Acetonitrile, anhydrous, 99.8%
C2H3N
675415
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
C2H3N
437557
Acetonitrile, ≥99.5%, ACS reagent
C2H3N
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, JIS 5000
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
639133
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
A6141
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
CH5NO3
D8284
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
C20H10N2Na2O4 · xH2O
SML0002
C646, ≥98% (HPLC)
C24H19N3O6
01810
Cycloheximide, ≥93.0% (HPLC)
C15H23NO4
C7698
Cycloheximide, from microbial, ≥94% (TLC)
C15H23NO4
C1988
Cycloheximide, Biotechnology Performance Certified
C15H23NO4
C4859
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, 0.2 μm filtered
C15H23NO4
11020
DL-Arginine, ≥97.0% (NT)
C6H14N4O2
EDS
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
C10H16N2O8