EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Large genomic fragment deletion and functional gene cassette knock-in via Cas9 protein mediated genome editing in one-cell rodent embryos.
PMID 26620761
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5770
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
C6H5Na3O7
83273
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
C6H5Na3O7
316989
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
H2O2
H1009
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
H2O2
40287
Hydrogen peroxide solution, semiconductor grade MOS PURANAL (Honeywell 17937), ≥30%
H2O2
18304
Hydrogen peroxide solution, meets analytical specification of Ph. Nord., 34.5-36.5%
H2O2
I6758
IPTG, ≥99% (TLC), ≤0.1% Dioxane
C9H18O5S
PHG0010
Isopropyl β-D-1-thiogalactopyranoside, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C9H18O5S
I5502
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
C9H18O5S
I1284
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
C9H18O5S
322415
Methanol, anhydrous, 99.8%
CH4O
M1770
Methanol, BioReagent, suitable for protein sequencing
CH4O
650609
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
CH4O
270474
Methanol, for HPLC, gradient grade, 99.93%
CH4O
65548
Methanol, for HPLC, gradient grade, ≥99.8% (GC)
CH4O
40217
Methanol, semiconductor grade PURANAL (Honeywell 17824)
CH4O
19-2470
Methanol, for HPLC
CH4O
19-2453
Methanol, for residue analysis, JIS 5000
CH4O
19-2400
Methanol, SAJ special grade
CH4O
19-2425
Methanol, suitable for determination of dioxins
CH4O
717975
Methanol, NMR reference standard
CH4O
650188
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
CH4O
632546
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, for mass spectrometry, ≥99.5%
CH4O
667390
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, for HPLC
CH4O
721964
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
CH4O
721956
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
CH4O
606723
Methanol-12C, 99.95 atom % 12C
12CH4O