EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Synthesis of novel pyridyl containing phospholanes and their polynuclear luminescent copper(i) complexes.
PMID 26621131
Abstract
A novel type of cyclic P,N-ligands, pyridyl containing phospholanes, has been synthesized in a moderate yield by the reaction of primary phosphines with 1,4-dichlorobutane in a superbasic medium. A series of homo tetranuclear octahedral Cu4I4L2, dinuclear tetrahedral Cu2I2L3, and dinuclear "head-to-tail" Cu2I2L2 luminescent complexes with these ligands were obtained. All the compounds were characterized using a range of spectroscopic and computational techniques, and in the case of some Cu4I4L2 and Cu2I2L3 complexes, by single crystal X-ray diffraction. The structural diversity of the obtained complexes was reflected in their photophysical properties: phosphorescence spectra of the compounds display emission in broad spectral range of 471-615 nm. TD-DFT computations allow the assignment of a single emission band around 550 nm for Cu2I2L3 complexes and 471 nm for Cu2I2L2 complex to a vertical triplet-singlet transition from a metal-to-ligand and halide-to-ligand charge-transfer (3)(M + X)LCT excited state, whereas a second band at around 600 nm in the spectra of octahedral Cu4I4L2 complexes was assigned predominantly to Cu4I4 cluster-centered ((3)CC) excited state.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131652
Pyridine N-oxide, 95%
C5H5NO