EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Jinmaitong decreases sciatic nerve DNA oxidative damage and apoptosis in a streptozotocin-induced diabetic rat model.
PMID 26622393
Abstract
Diabetic peripheral neuropathy (DPN) is a common chronic complication of diabetes. Jinmaitong (JMT), a Traditional Chinese Medicine, improves certain symptoms of DPN, such as limb pain and numbness. The aim of the present study was to investigate the effects of JMT on DNA oxidative damage and apoptosis in the sciatic nerve of diabetic rats. The rats were divided into a normal and a diabetic group. Diabetes was induced using streptozotocin (60 mg/kg). The diabetic model (DM) rats received vitamin C (0.05 g/kg/day) or JMT [low-dosage (L), 0.44 g/kg/day; medium-dosage (M), 0.88 g/kg/day or high-dosage (H), 1.75 g/kg/day]. After 16 weeks, the mechanical pain threshold of the rats was evaluated. The expression of 8-hydroxy-deoxyguanosine (8-OHdG), nicotinamide adenine dinucleotide phosphate (NADPH) oxidase p22