EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Effect of serum from postmenopausal women with osteoporosis exhibiting the Kidney-Yang deficiency pattern on bone formation in an hFOB 1.19 human osteoblastic cell line.
PMID 26622445
Abstract
The aim of the present study was to investigate the underlying mechanism of the Kidney-Yang deficiency (KYD) pattern of osteoporosis in postmenopausal women of a certain age range by comparing the effect of serum from postmenopausal women with osteoporosis exhibiting the KYD pattern with that of serum from postmenopausal women without osteoporosis on bone formation in an hFOB 1.19 human osteoblastic cell line. A random selection of 30 female, postmenopausal volunteers aged 60-70 years, including 15 cases without osteoporosis and 15 cases with the KYD pattern of osteoporosis, were enrolled at the Physical Examination Center of the Second Affiliated Hospital of Fujian University of Traditional Chinese Medicine. Venous blood was extracted and the serum was separated. The hFOB 1.19 cells were treated with 10% KYD pattern-serum or control serum from postmenopausal women of the same age range without osteoporosis. It was found that the KYD pattern-serum significantly decreased the cell viability, activity of alkaline phosphatase and number of calcified nodules, as well as downregulated the expression of osteocalcin and osteoprotegerin (OPG) and upregulated that of receptor activator of nuclear factor κB ligand (RANKL) in the hFOB 1.19 cells. In addition, the present results showed that the concentrations of estradiol (E