EMAIL THIS PAGE TO A FRIEND
Oncology letters
Guggulsterone inhibits human cholangiocarcinoma Sk-ChA-1 and Mz-ChA-1 cell growth by inducing caspase-dependent apoptosis and downregulation of survivin and Bcl-2 expression.
PMID 26622683
Abstract
Guggulsterone has recently been reported to demonstrate anti-tumor effects in a variety of cancers. The present study aims to investigate the biological roles and underlying mechanism of the action of guggulsterone in cholangiocarcinoma. The immortalized human cholangiocarcinoma Sk-ChA-1 and Mz-ChA-1 cell lines were treated with various concentrations of the