PLoS ONE
Loss of Gadkin Affects Dendritic Cell Migration In Vitro.
PMID 26624014
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
H4034
HEPES, BioPerformance Certified, ≥99.5% (titration), cell culture tested
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
RES6003H-B7
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
H7523
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
C8H18N2O4S
H6147
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
C8H18N2O4S
54457
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
C8H18N2O4S
83264
HEPES buffer solution, 1 M in H2O
C8H18N2O4S
G3126
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
C5H10N2O3
G5792
L-Glutamine, PharmaGrade, Ajinomoto, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H10N2O3
G8540
L-Glutamine, meets USP testing specifications, cell culture tested, 99.0-101.0%, from non-animal source
C5H10N2O3
G6392
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
C5H10N2O3
49419
L-Glutamine, BioUltra, ≥99.5% (NT)
C5H10N2O3
W368401
L-Glutamine
C5H10N2O3
P7626
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
C7H7FO2S
78830
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
C7H7FO2S
746436
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
ClK
793590
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
ClK
P9333
Potassium chloride, BioXtra, ≥99.0%
ClK
P9541
Potassium chloride, for molecular biology, ≥99.0%
ClK
P5405
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
ClK
P1597
Potassium chloride, meets EP, USP testing specifications
ClK
449989
Potassium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClK
451029
Potassium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
ClK
409316
Potassium chloride, 99.999% trace metals basis
ClK
204099
Potassium chloride, ≥99.99% trace metals basis
ClK
60128
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClK
P9327
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
ClK
60142
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
ClK