EMAIL THIS PAGE TO A FRIEND
Nanoscale Research Letters
Enhanced Electrochemical Catalytic Efficiencies of Electrochemically Deposited Platinum Nanocubes as a Counter Electrode for Dye-Sensitized Solar Cells.
PMID 26625891
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9516
2-Propanol, BioReagent, for molecular biology, ≥99.5%
C3H8O
278475
2-Propanol, anhydrous, 99.5%
C3H8O
675431
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
C3H8O
437522
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59304
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C3H8O
40301
2-Propanol, semiconductor grade VLSI PURANAL (Honeywell 17604)
C3H8O
40219
2-Propanol, semiconductor grade PURANAL (Honeywell 17829)
C3H8O
270490
2-Propanol, 99.5%, HPLC grade
C3H8O
15-2360
2-Propanol, for HPLC
C3H8O
733458
2-Propanol, electronic grade, 99.999% trace metals basis
C3H8O
439126
Acetone, for HPLC, ≥99.9%
C3H6O
00561
Acetone, for luminescence, ≥99.5% (GC)
C3H6O
40289
Acetone, semiconductor grade VLSI PURANAL (Honeywell 17617)
C3H6O
W332607
Acetone, ≥99%, meets FCC analytical specifications
C3H6O
W332615
Acetone, natural, ≥97%
C3H6O
01-0520
Acetone, for HPLC
C3H6O
01-0494
Acetone, for residue analysis, JIS 5000
C3H6O
01-0475
Acetone, suitable for determination of dioxins
C3H6O
48358
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
C3H6O
686530
Acetone solution, contains 20.0 % (v/v) acetonitrile, for HPLC
C3H6O
W292907
Isopropyl alcohol, ≥99.7%, FCC, FG
C3H8O