EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Collaborative regulation of CO2 transport and fixation during succinate production in Escherichia coli.
PMID 26626308
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
271004
Acetonitrile, anhydrous, 99.8%
C2H3N
675415
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
C2H3N
437557
Acetonitrile, ≥99.5%, ACS reagent
C2H3N
A3396
Acetonitrile, ≥99.5%
C2H3N
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, JIS 5000
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
639133
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
449172
Magnesium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl2Mg
449164
Magnesium chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Mg
M8266
Magnesium chloride, anhydrous, ≥98%
Cl2Mg
M4880
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Cl2Mg
208337
Magnesium chloride, powder, <200 μm
Cl2Mg
M1028
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Cl2Mg
63069
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Cl2Mg
M8787
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Cl2Mg
63036
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Cl2Mg
68475
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Cl2Mg
00457
Magnesium chloride solution, 0.1 M
Cl2Mg