EMAIL THIS PAGE TO A FRIEND
Scientific reports
Tunable-Porosity Membranes From Discrete Nanoparticles.
PMID 26626565
Abstract
Thin film composite membranes were prepared through a facile single-step wire-wound rod coating procedure in which internally crosslinked poly(styrene-co-butadiene) polymer nanoparticles self-assembled to form a thin film on a hydrophilic ultrafiltration support. This nanoparticle film provided a defect-free separation layer 130-150 nm thick, which was highly permeable and able to withstand aggressive pH conditions beyond the range of available commercial membranes. The nanoparticles were found to coalesce to form a rubbery film when heated above their glass transition temperature (Tg). The retention properties of the novel membrane were strongly affected by charge repulsion, due to the negative charge of the hydroxyl functionalized nanoparticles. Porosity was tuned by annealing the membranes at different temperatures, below and above the nanoparticle Tg. This enabled fabrication of membranes with varying performance. Nanofiltration properties were achieved with a molecular weight cut-off below 500 g mol(-1) and a low fouling tendency. Interestingly, after annealing above Tg, memory of the interstitial spaces between the nanoparticles persisted. This memory led to significant water permeance, in marked contrast to the almost impermeable films cast from a solution of the same polymer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
746452
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
MgSO4
M2643
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
MgSO4
793612
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
MgSO4
203726
Magnesium sulfate, ≥99.99% trace metals basis
MgSO4
83266
Magnesium sulfate solution, BioUltra, for molecular biology
MgSO4
M3409
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
MgSO4
227056
N,N-Dimethylformamide, anhydrous, 99.8%
C3H7NO
D4551
N,N-Dimethylformamide, for molecular biology, ≥99%
C3H7NO
648531
N,N-Dimethylformamide, for HPLC, ≥99.9%
C3H7NO
437573
N,N-Dimethylformamide, ACS reagent, ≥99.8%
C3H7NO
S9000
Sodium metabisulfite, ReagentPlus®, ≥99%
Na2O5S2
31448
Sodium metabisulfite, puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%
Na2O5S2
13459
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Na2O5S2
161519
Sodium metabisulfite, reagent grade, 97%
Na2O5S2
799343
Sodium metabisulfite, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Na2O5S2
255556
Sodium metabisulfite, ACS reagent, ≥97.0%
Na2O5S2
S4972
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
C8H8
47745-U
Styrene, analytical standard
C8H8
48347
Styrene solution, certified reference material, 200 μg/mL in methanol
C8H8
244511
Toluene, anhydrous, 99.8%
C7H8
648566
Toluene, for HPLC, ≥99.9%
C7H8
270377
Toluene, HPLC grade, 99.8%
C7H8
40248
N,N-Dimethylformamide, puriss., absolute, over molecular sieve (H2O ≤0.01%), ≥99.8% (GC)
C3H7NO