EMAIL THIS PAGE TO A FRIEND
Scientific reports
Discovery of a Novel Inhibitor of the Protein Tyrosine Phosphatase Shp2.
PMID 26626996
Abstract
Shp2 is a ubiquitously expressed protein tyrosine phosphatase (PTP) related to adult acute myelogenous leukemia and human solid tumors. In this report, we describe identification of a potent Shp2 inhibitor, Fumosorinone (Fumos) from entomogenous fungi, which shows selective inhibition of Shp2 over other tested PTPs. Using a surface plasmon resonance analysis, we further confirmed the physical interaction between Shp2 and Fumos. Fumos inhibits Shp2-dependent activation of the Ras/ERK signal pathway downstream of EGFR, and interrupts EGF-induced Gab1-Shp2 association. As expected, Fumos shows little effects on the Shp2-independent ERK1/2 activation induced by PMA or oncogenic Ras. Furthermore, Fumos down-regulates Src activation, inhibits phosphorylation of Paxillin and prevents tumor cell invasion. These results suggest that Fumos can inhibit Shp2-dependent cell signaling in human cells and has a potential for treatment of Shp2-associated diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43816
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
C4H10O2S2
646563
DL-Dithiothreitol solution, 1 M in H2O
C4H10O2S2
G2289
Glycerin, meets USP testing specifications
C3H8O3
G5516
Glycerol, for molecular biology, ≥99%
C3H8O3
G6279
Glycerol, BioXtra, ≥99% (GC)
C3H8O3
G9012
Glycerol, ≥99.5%
C3H8O3
G2025
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
C3H8O3
49767
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
C3H8O3
W252506
Glycerol, >99%, FCC, FG
C3H8O3
49781
Glycerol solution, 86-89% (T)
C3H8O3 · xH2O
792527
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
C3H4N2
I5513
Imidazole, for molecular biology, ≥99% (titration)
C3H4N2
I0250
Imidazole, ≥99% (titration), crystalline
C3H4N2
I2399
Imidazole, ACS reagent, ≥99% (titration)
C3H4N2
56750
Imidazole, puriss. p.a., ≥99.5% (GC)
C3H4N2
I202
Imidazole, ReagentPlus®, 99%
C3H4N2
56748
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
C3H4N2
56749
Imidazole, BioUltra, ≥99.5% (GC)
C3H4N2
68268
Imidazole buffer Solution, BioUltra, 1 M in H2O
C3H4N2
322415
Methanol, anhydrous, 99.8%
CH4O
650609
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
CH4O
270474
Methanol, for HPLC, gradient grade, 99.93%
CH4O
65548
Methanol, for HPLC, gradient grade, ≥99.8% (GC)
CH4O
19-2470
Methanol, for HPLC
CH4O
19-2453
Methanol, for residue analysis, JIS 5000
CH4O
19-2400
Methanol, SAJ special grade
CH4O
19-2425
Methanol, suitable for determination of dioxins
CH4O
717975
Methanol, NMR reference standard
CH4O
650188
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
CH4O
632546
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, for mass spectrometry, ≥99.5%
CH4O