EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
A meningococcal vaccine antigen engineered to increase thermal stability and stabilize protective epitopes.
PMID 26627237
Abstract
Factor H binding protein (FHbp) is part of two vaccines recently licensed for prevention of sepsis and meningitis caused by serogroup B meningococci. FHbp is classified in three phylogenic variant groups that have limited antigenic cross-reactivity, and FHbp variants in one of the groups have low thermal stability. In the present study, we replaced two amino acid residues, R130 and D133, in a stable FHbp variant with their counterparts (L and G) from a less stable variant. The single and double mutants decreased thermal stability of the amino- (N-) terminal domain compared with the wild-type protein as measured by scanning calorimetry. We introduced the converse substitutions, L130R and G133D, in a less stable wild-type FHbp variant, which increased the transition midpoint (Tm) for the N-terminal domain by 8 and 12 °C; together the substitutions increased the Tm by 21 °C. We determined the crystal structure of the double mutant FHbp to 1.6 Å resolution, which showed that R130 and D133 mediated multiple electrostatic interactions. Monoclonal antibodies specific for FHbp epitopes in the N-terminal domain had higher binding affinity for the recombinant double mutant by surface plasmon resonance and to the mutant expressed on meningococci by flow cytometry. The double mutant also had decreased binding of human complement Factor H, which in previous studies increased the protective antibody responses. The stabilized mutant FHbp thus has the potential to stabilize protective epitopes and increase the protective antibody responses to recombinant FHbp vaccines or native outer membrane vesicle vaccines with overexpressed FHbp.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8032
Sodium azide, BioXtra
N3Na
438456
Sodium azide, ≥99.99% trace metals basis
N3Na
S2002
Sodium azide, ReagentPlus®, ≥99.5%
N3Na
71289
Sodium azide, BioUltra, ≥99.5% (T)
N3Na
71290
Sodium azide, purum p.a., ≥99.0% (T)
N3Na
RES0926S-A7
Sodium chloride, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
ClNa
S7653
Sodium chloride, BioXtra, ≥99.5% (AT)
ClNa
S3014
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥98% (titration)
ClNa
S5886
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
ClNa
S1679
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
ClNa
S6150
Sodium chloride, tablet
ClNa
S6191
Sodium chloride, BioPerformance Certified, ≥99% (titration), Cell Culture Tested
ClNa
450006
Sodium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClNa
450014
Sodium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
ClNa
204439
Sodium chloride, 99.999% trace metals basis
ClNa
378860
Sodium chloride, random crystals, optical grade, 99.98% trace metals basis
ClNa
310166
Sodium chloride, reagent grade, ≥98%, +80 mesh particle size
ClNa
71376
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClNa
28-2270
Sodium chloride, JIS special grade, ≥99.5%
ClNa
S5150
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
ClNa
S6546
Sodium chloride solution, 5 M
ClNa
59222C
Sodium chloride solution, 5 M
ClNa
71386
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
ClNa
S0817
Sodium chloride solution, 0.85%
ClNa
16818
Sodium chloride solution, 2 M
ClNa
S8776
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
ClNa
451940
Sodium chloride-35Cl, 99 atom % 35Cl
35ClNa