EMAIL THIS PAGE TO A FRIEND
Microbial cell factories
Deletion of the 2-acyl-glycerophosphoethanolamine cycle improve glucose metabolism in Escherichia coli strains employed for overproduction of aromatic compounds.
PMID 26627477