EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Human embryonic stem cell-derived mesenchymal cells preserve kidney function and extend lifespan in NZB/W F1 mouse model of lupus nephritis.
PMID 26628350