EMAIL THIS PAGE TO A FRIEND
International Journal of Clinical and Experimental Medicine
Genetic association analysis between polymorphisms of HAIRY-AND-ENHANCER-OF SPLIT-7 and congenital scoliosis.
PMID 26629209
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30692
Decylamine, ≥99.0% (GC)
C10H23N
D2404
Decylamine, 95%
C10H23N
442540
Decylamine, analytical standard
C10H23N