EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental medicine
Resveratrol induces apoptosis in K562 cells via the regulation of mitochondrial signaling pathways.
PMID 26629245
Abstract
Resveratrol, an edible polyphenolic phytoalexin obtained primarily from root extracts of the oriental plant, Polygonum cuspidatum and from grapes and red wine, has been reported as an anticancer compound against several types of cancer, the accurate molecular mechanisms of by which it induces apoptosis are limited. In the present study, the molecular mechanisms of resveratrol on human leukemia K562 cells apoptosis was examined. Our results showed that resveratrol significantly decreased cell viability and triggered cell apoptosis in K562 cells. Resveratrol-induced apoptosis of K562 cells was associated with the dissipation of mitochondrial membrane potential (MMP) and the release of cytochrome c into the cytosol. Furthermore, the up-regulation of Bax/Bcl-2 ratio, the activation of caspase-3 and increased cleaved PARP was also observed in K562 cells treated with resveratrol. Thus, we considered that the resveratrol-induced apoptosis of K562 cells might be mediated through the mitochondria pathway, which gives the rationale for in vivo studies on the utilization of resveratrol as a potential cancer therapeutic compound.