PLoS ONE
Dystroglycan Depletion Impairs Actin-Dependent Functions of Differentiated Kasumi-1 Cells.
PMID 26630171
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74875
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
C24H21Na3O11S3
C8273
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
C30H37NO6
C2618
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO, 0.2 μm filtered
C30H37NO6
D2650
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
C2H6OS
D8418
Dimethyl sulfoxide, for molecular biology
C2H6OS
D4540
Dimethyl sulfoxide, ≥99.5% (GC), plant cell culture tested
C2H6OS
D1435
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
C2H6OS
D9170
Dimethyl sulfoxide, PCR Reagent
C2H6OS
D2438
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
C2H6OS
276855
Dimethyl sulfoxide, anhydrous, ≥99.9%
C2H6OS
41639
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
C2H6OS
W387509
Dimethyl sulfoxide, ≥99%
C2H6OS
07-4860
Dimethyl sulfoxide, JIS special grade, ≥99.0%
C2H6OS
07-4871
Dimethyl sulfoxide, for HPLC
C2H6OS
673439 Dimethyl sulfoxide solution, 50 wt. % in H2O
C2H6OS
606987
Dimethyl sulfoxide-12C2, 99.95 atom % 12C
C2H6SO
L2020 Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris buffered NaCl, 0.2 μm filtered, BioReagent, suitable for cell culture