EMAIL THIS PAGE TO A FRIEND
Scientific reports
Flower-like PEGylated MoS2 nanoflakes for near-infrared photothermal cancer therapy.
PMID 26632249
Abstract
Photothermal cancer therapy has attracted considerable interest for cancer treatment in recent years, but the effective photothermal agents remain to be explored before this strategy can be applied clinically. In this study, we therefore develop flower-like molybdenum disulfide (MoS2) nanoflakes and investigate their potential for photothermal ablation of cancer cells. MoS2 nanoflakes are synthesized via a facile hydrothermal method and then modified with lipoic acid-terminated polyethylene glycol (LA-PEG), endowing the obtained nanoflakes with high colloidal stability and very low cytotoxicity. Upon irradiation with near infrared (NIR) laser at 808 nm, the nanoflakes showed powerful ability of inducing higher temperature, good photothermal stability and high photothermal conversion efficiency. The in vitro photothermal effects of MoS2-PEG nanoflakes with different concentrations were also evaluated under various power densities of NIR 808-nm laser irradiation, and the results indicated that an effective photothermal killing of cancer cells could be achieved by a low concentration of nanoflakes under a low power NIR 808-nm laser irradiation. Furthermore, cancer cell in vivo could be efficiently destroyed via the photothermal effect of MoS2-PEG nanoflakes under the irradiation. These results thus suggest that the MoS2-PEG nanoflakes would be as promising photothermal agents for future photothermal cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323446
Ammonium tetrathiomolybdate, 99.97% trace metals basis
H8MoN2S4
379115
DCC, 1.0 M in methylene chloride
C13H22N2
D80002
DCC, 99%
C13H22N2
633267
Dicyclohexylcarbodiimide solution, 60 wt. % in xylenes
C13H22N2
H9627
Hematoxylin
C16H14O6 · xH2O
H3136
Hematoxylin, certified by the Biological Stain Commission
C16H14O6 · xH2O
225819
Hydrazine hydrate, reagent grade, N2H4 50-60 %
H4N2 · xH2O
53847
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)
H4N2 · H2O
15622
Hydrazine hydrate solution, 24-26% (iodometric)
H4N2 · H2O
18412
Hydrazine hydrate solution, 78-82% (iodometric)
H4N2 · H2O
207942
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, 98%
H4N2 · H2O
T0886
Triethylamine, ≥99%
C6H15N
90335
Triethylamine, BioUltra, ≥99.5% (GC)
C6H15N
90337
Triethylamine, for amino acid analysis, ≥99.5% (GC)
C6H15N
90338
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
C6H15N
90340
Triethylamine, puriss. p.a., ≥99.5% (GC)
C6H15N
16304
Triethylamine, purum, ≥99% (GC)
C6H15N
471283
Triethylamine, ≥99.5%
C6H15N
W424601
Triethylamine, ≥99.5%
C6H15N
243205
Trimethylamine, anhydrous, ≥99%
C3H9N
92260
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
C3H9N
92262
Trimethylamine solution, ~45 wt. % in H2O (T)
C3H9N
W324108
Trimethylamine solution, 25 wt. % in H2O
C3H9N
W550132
Trimethylamine solution, 25 wt. % in propylene glycol
C3H9N
302643
Trypan Blue, Dye content 60 %
C34H24N6Na4O14S4
T6146
Trypan Blue, powder, BioReagent, suitable for cell culture
C34H24N6Na4O14S4
T8154
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
C34H24N6Na4O14S4