EMAIL THIS PAGE TO A FRIEND
Scientific reports
MicroRNA-195 inhibits proliferation, invasion and metastasis in breast cancer cells by targeting FASN, HMGCR, ACACA and CYP27B1.
PMID 26632252
Abstract
De novo lipogenesis, a hallmark for cancers is required for cellular transformation. Further it is believed that resistance to apoptosis and epithelial-to-mesenchymal-transition(EMT) facilitates metastasis via over-expression of anti-apoptotic Bcl-2. Previously we demonstrated that hsa-miR-195 targets BCL2, induces apoptosis and augmented the effect of etoposide in breast cancer cells. However, the mechanism behind its function remains elusive. Herein gene expression profiling was done in presence/absence of hsa-miR-195 in Breast cancer cells. IPA revealed mitochondrial dysfunction, fatty acid metabolism and xenobiotic metabolism signalling among the top processes being affected. For the first time we herein identified ACACA, FASN (the key enzymes of de novo fatty acid synthesis), HMGCR (the key enzyme of de novo cholesterol synthesis) and CYP27B1 as direct targets of hsa-miR-195. We further showed that ectopic expression of hsa-miR-195 in MCF-7 and MDA-MB-231 cells not only altered cellular cholesterol and triglyceride levels significantly but also resulted in reduced proliferation, invasion and migration. We further demonstrated that over expression of hsa-miR-195 decreased the Mesenchymal markers expression and enhanced Epithelial markers. In conclusion we say that hsa-miR-195 targets the genes of de novo lipogenesis, inhibits cell proliferation, migration, and invasion which potentially opens new avenues for the treatment of breast cancer.