EMAIL THIS PAGE TO A FRIEND
Scientific reports
Comparing photosynthetic characteristics of Isoetes sinensis Palmer under submerged and terrestrial conditions.
PMID 26634994
Abstract
Crassulacean acid metabolism (CAM) is widespread in terrestrial and aquatic species, plastic in response to environmental changes. Isoetes L. is one of the earliest basal vascular plants and CAM is popular in this genus. Isoetes sinensis Palmer is an amphibious species, alternating frequently between terrestrial and aquatic environments. Given this, we investigated and compared photosynthetic characteristics over a diurnal cycle under submerged condition (SC) and terrestrial condition (TC). The results suggest that I. sinensis possesses a stronger CAM capacity under SC. Compared with under TC, titratable acidity levels and organic acid concentrations were more enriched under SC, whereas soluble sugar or starch and protein levels were lower under SC. Transcript analyses for nine photosynthetic genes revealed that CAM-associated genes possessed high transcripts under SC, but C3-related transcripts were highly expressed under TC. In addition, the enzyme activity measurements demonstrated that PEPC activity over a diurnal cycle was slightly higher under SC, whereas Rubisco activity during the daytime was greater under TC. This comprehensive study probably facilitates general understandings about the CAM photosynthetic characteristics of Isoetes in response to the environmental changes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43816
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
C4H10O2S2
646563
DL-Dithiothreitol solution, 1 M in H2O
C4H10O2S2
E9884
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
C10H16N2O8
EDS
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
C10H16N2O8
E6758
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
C10H16N2O8
431788
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
C10H16N2O8
ED
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
C10H16N2O8
E8008
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
C10H16N2O8
M8266
Magnesium chloride, anhydrous, ≥98%
Cl2Mg
208337
Magnesium chloride, powder, <200 μm
Cl2Mg
449172
Magnesium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl2Mg
M4880
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Cl2Mg
449164
Magnesium chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Mg
M1028
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Cl2Mg
63069
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Cl2Mg
M8787
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Cl2Mg
68475
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Cl2Mg
00457
Magnesium chloride solution, 0.1 M
Cl2Mg
63036
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Cl2Mg