EMAIL THIS PAGE TO A FRIEND
Frontiers in Plant Science
Global analysis of WRKY transcription factor superfamily in Setaria identifies potential candidates involved in abiotic stress signaling.
PMID 26635818