PloS one
Identification and Expression Analysis of a Novel HbCIPK2-Interacting Ferredoxin from Halophyte H. brevisubulatum.
PMID 26636581
Abstract
Ferredoxin is a small iron-sulfer protein involved in various one-eletron transfer pathways. Little is known about how ferredoxin is regulated to distribute electron under abiotic stress. Our previous study has showed that HbCIPK2 conferred salinity and drought tolerance. Thus, we hypothesized that HbCIPK2 could mediate the activities of interacting partners as a signal transducer. In this report, we identified a novel HbCIPK2-interacting ferredoxin (HbFd1) from halophyte Hordeum brevisubulatum by yeast two-hybrid screens, confirmed this interaction by BiFC in vivo and CoIP in vitro, and presented the expression pattern of HbFd1. HbFd1 was down-regulated under salinity and cold stress but up-regulated under PEG stress, its expression showed tissue-specific, mainly in shoot chloroplast, belonging to leaf-type subgroup. Moreover, HbCIPK2 could recruit HbFd1 to the nucleus for their interaction. The C-terminal segment in HbFd1 protein was involved in the interaction with HbCIPK2. These results provided insight into the connection between CBL-CIPK signaling network and Fd-dependent metabolic pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9546
D-Mannitol, BioXtra, ≥98% (HPLC)
C6H14O6
M9647
D-Mannitol, ACS reagent
C6H14O6
M4125
D-Mannitol, ≥98%
C6H14O6
M1902
D-Mannitol, plant cell culture tested
C6H14O6
M8429
D-Mannitol, meets EP, FCC, USP testing specifications
C6H14O6
63559
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C6H14O6
RES0926S-A7
Sodium chloride, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
ClNa
S7653
Sodium chloride, BioXtra, ≥99.5% (AT)
ClNa
S3014
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥98% (titration)
ClNa
S5886
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
ClNa
S1679
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
ClNa
S6150
Sodium chloride, tablet
ClNa
S6191
Sodium chloride, BioPerformance Certified, ≥99% (titration), Cell Culture Tested
ClNa
450006
Sodium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClNa
204439
Sodium chloride, 99.999% trace metals basis
ClNa
378860
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
ClNa
71376
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClNa
S5150
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
ClNa
S6546
Sodium chloride solution, 5 M
ClNa
59222C
Sodium chloride solution, 5 M
ClNa
71386
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
ClNa
S0817
Sodium chloride solution, 0.85%
ClNa
S8776
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
ClNa
451940
Sodium chloride-35Cl, 99 atom % 35Cl
35ClNa