EMAIL THIS PAGE TO A FRIEND
Journal of Cardiothoracic Surgery
Six-years survival and predictors of mortality after CABG using cold vs. warm blood cardioplegia in elective and emergent settings.
PMID 26637200