EMAIL THIS PAGE TO A FRIEND
Scientific Reports
RESOLFT nanoscopy with photoswitchable organic fluorophores.
PMID 26639557
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5262 Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
D22606
Cadaverine, 95%
C5H14N2
G2133
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
L4170
LDN-57444, ≥98% (HPLC)
C17H11Cl3N2O3
P6148
Paraformaldehyde, reagent grade, crystalline
CH2O ]n
10981 Polyvinyl alcohol mounting medium with DABCO®, antifading, pH 8.7
71320
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
H4BNa
452890
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
H4BNa
452165
Sodium borohydride, caplets
H4BNa
480886
Sodium borohydride, granular, 99.99% trace metals basis
H4BNa
452874
Sodium borohydride, granular, 10-40 mesh, 98%
H4BNa
452882
Sodium borohydride, powder, ≥98.0%
H4BNa
247677
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
H4BNa
71318
Sodium borohydride, purum, ≥97.0% (gas-volumetric), tablet
H4BNa
213462
Sodium borohydride, ReagentPlus®, 99%
H4BNa
686018
Sodium borohydride, hydrogen-storage grade, 98%
H4BNa
200972
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
H4BNa
452904
Sodium borohydride solution, SWS, ~12 wt. % in 14 M NaOH
H4BNa
215511
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
H4BNa
T0886
Triethylamine, ≥99%
C6H15N
90335
Triethylamine, BioUltra, ≥99.5% (GC)
C6H15N
90337
Triethylamine, for amino acid analysis, ≥99.5% (GC)
C6H15N
90338
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
C6H15N
90340
Triethylamine, puriss. p.a., ≥99.5% (GC)
C6H15N
16304
Triethylamine, purum, ≥99% (GC)
C6H15N
471283
Triethylamine, ≥99.5%
C6H15N
W424601
Triethylamine, ≥99.5%
C6H15N
T8787
Triton X-100, for molecular biology
632287
VenPure® SF, powder
H4BNa