EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Thymocytes maintain immune activity through telomere elongation in rats under hypoxic conditions.
PMID 26640565
Abstract
The main purpose of the present study was to investigate the change in thymocyte telomere length of rats exposed to different hypoxic conditions for different periods of time, as well as its effect on the immune system. A total of 110 male Wistar rats were randomly assigned to one of the three following groups: i) Sea level (SL) group, in which 10 rats were maintained at an altitude of 10 m; ii) moderate altitude (MA) group, in which 50 rats were maintained at an altitude of 2,260 m and then randomly sacrificed on days 1, 3, 7, 15 and 30 (n=10 each); and iii) simulated high altitude (SHA) group, in which 50 rats were maintained at a simulated altitude of 5,000 m, and then randomly sacrificed on days 1, 3, 7, 15 and 30 (n=10 each). The morphological changes of the thymus were observed, while the telomere length, the mRNA and protein expression levels of telomerase reverse transcriptase (TERT), and the peripheral blood lymphocyte count were measured. The results indicated that hypoxia induced morphological changes and apoptosis in thymocytes, as well as atrophy of the thymus tissue, and resulted in a significant increase in telomere length and TERT mRNA and protein expression levels. This effect appeared to be more pronounced in the SHA group compared with that in the MA group; however, no statistically significant changes were observed in the peripheral blood lymphocyte count. Based on these findings, the hypoxia-associated loss of thymic function appears to be only quantitative and not qualitative, and the thymus may be able to maintain its immune function even under hypoxic conditions.