EMAIL THIS PAGE TO A FRIEND
Nanoscale research letters
Manganese(II) Complexes with Schiff Bases Immobilized on Nanosilica as Catalysts of the Reaction of Ozone Decomposition.
PMID 26643653
Abstract
In this article, we submit the description of synthesis and identification of manganese(II) complexes with pyrogenic nanosilica-immobilized (d av = 10 nm; S sp = 290 m(2)/g) hydroxyaldimine ligands [Formula: see text]: salicilaldiminopropyl (L1); 5-bromosalicilaldiminopropyl (L2); 2-hydroxynaphtaldiminopropyl (L3); 2-hydroxy-3-methoxybenzaldiminopropyl (L4); 2-hydroxy-3,5-dichloroacetophenoniminopropyl (L5); and 4-hydroxy-3-methoxybenzaldiminopropyl (L6). The ligands and complexes were characterized by UV-VIS and IR spectrometry. Nanocomposites consisting of complexes [Formula: see text] showed a high catalytic activity in low-temperature ozone decomposition in the range of concentrations between 2.1 × 10(-6) and 8.4 × 10(-6) mol/l. The number of catalytic cycles increased for isostructural pseudotetrahedral complexes [Formula: see text] (L1-L5) in the following order: Mn(L3)2 > Mn(L4)2 > Mn(L1)2 > Mn(L2)2 > Mn(L5)2. In the case of pseudooctahedral complexes with L6, the change of coordination polyhedral does not influence the kinetics and stoichiometric parameters of the reaction.