EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Effects of Experimental Coma on the Expression of Bcl-2 Protein and Caspases 3 and 9 in Rat Brain.
PMID 26645287
Abstract
We performed immunohistochemical analysis of the expression of caspases 3, 9 and bcl-2 protein in rat brain at various terms after administration of LD50 of sodium thiopental. Expression of the specified apoptosis markers was found in the sensorimotor cortex and hippocampus (dentate gyrus and CA2 region).