EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Studies on the biosynthesis of ralfuranones in Ralstonia solanacearum.
PMID 26645956
Abstract
Ralfuranones, aryl-furanone secondary metabolites, are involved in the virulence of Ralstonia solanacearum in solanaceous plants. Ralfuranone I (6) has been suggested as a biosynthetic precursor for other ralfuranones; however, this conversion has not been confirmed. We herein investigate the biosynthesis of ralfuranones using feeding experiments with ralfuranone I (6) and its putative metabolite, ralfuranone B (2). The results obtained demonstrated that the biosynthesis of ralfuranones proceeded in enzymatic and non-enzymatic manners.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
XAD16 Amberlite® XAD16N, 20-60 mesh