EMAIL THIS PAGE TO A FRIEND
Lab on a chip
Highly focused high-frequency travelling surface acoustic waves (SAW) for rapid single-particle sorting.
PMID 26646200
Abstract
High-speed sorting is an essential process in a number of clinical and research applications, where single cells, droplets and particles are segregated based on their properties in a continuous flow. With recent developments in the field of microscale actuation, there is increasing interest in replicating the functions available to conventional fluorescence activated cell sorting (FACS) flow cytometry in integrated on-chip systems, which have substantial advantages in cost and portability. Surface acoustic wave (SAW) devices are ideal for many acoustofluidic applications, and have been used to perform such sorting at rates on the order of kHz. Essential to the accuracy of this sorting, however, is the dimensions of the region over which sorting occurs, where a smaller sorting region can largely avoid inaccurate sorting across a range of sample concentrations. Here we demonstrate the use of flow focusing and a highly focused SAW generated by a high-frequency (386 MHz), 10 μm wavelength set of focused interdigital transducers (FIDTs) on a piezoelectric lithium niobate substrate, yielding an effective sorting region only ~25 μm wide, with sub-millisecond pulses generated at up to kHz rates. Furthermore, because of the use of high frequencies, actuation of particles as small as 2 μm can be realized. Such devices represent a substantial step forward in the evolution of highly localized forces for lab-on-a-chip microfluidic applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
214698
Pyridinium dichromate, 98%
C10H10N2 · H2Cr2O7