EMAIL THIS PAGE TO A FRIEND
Acta cirurgica brasileira
Effect of alpha-7 nicotinic acetylcholine receptor activation on beta-amyloid induced recognition memory impairment. Possible role of neurovascular function.
PMID 26647792
Abstract
To evaluate the effects of PHA-543613 (α7-nAChR agonist) and galantamine (acetylcholinesterase inhibitor (AChEI)) on recognition memory and neurovascular coupling (NVC) response in beta-amyloid (Aβ) 25-35-treated mice. PHA-543613 (1 mg/kg, i.p.), and galantamine (3 mg/kg, s.c.), effects were tested in Aβ25-35 mice model of AD. α7-nAChR antagonist, methyllycaconitine (MLA) (1 mg/kg, i.p.), was used for evaluation of receptor blockade effects. Recognition memory in animals was assessed by the novel object recognition (NOR) task. NVC response was analyzed by laser-doppler flow meter in barrel cortex by whisker stimulation method. Both, PHA-543613 and galantamine improve recognition memory in Aβ-treated animals. However, the advantageous effects of PHA-543613 were significantly higher than galantamine. Also, pretreatment with MLA reversed both galantamine and PHA-543613 effects on NOR. Impaired NVC response in AD animals was improved by PHA-543613 and galantamine. However, MLA pretreatment disrupts this function. Activation of α7-nAChR improved recognition memory possible through enhancement of neurovascular response in Alzheimer's disease in animals.